Orders LinkContactUsLinkComponent
Iris scissors
Iris scissors
Straight, sharp/sharp, TC Gold, 115 mm
Catalog # MART11-982-11
Supplier:   KLS Martin Group
Iris scissors
Catalog # MART11-982-11
Supplier:  KLS Martin Group
Iris scissors
Straight, sharp/sharp, TC Gold, 115 mm
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Dissecting scissors made of stainless steel. One model features a TC Gold finish.
  • TC Gold finish: Tungsten Carbide inlays provide an exceptionally long service life
  • Straight or curved available
Specifications
  • Type:
    Straight, sharp/sharp, TC Gold
  • Length:
    115 mm
  • Seq:
    02