Orders LinkContactUsLinkComponent
Accessories for parallel synthesis, Syncore® Polyvap, Syncore® Analyst
Accessories for parallel synthesis, Syncore® Polyvap, Syncore® Analyst
Glass sample tubes for rack R-12 polyvap, round bottom (Ø 48 mm), height 170 mm, for working volume up to 120 ml
Catalog # BUCH040907
Supplier:   BUCHI
Accessories for parallel synthesis, Syncore® Polyvap, Syncore® Analyst
Catalog # BUCH040907
Supplier:  BUCHI
Accessories for parallel synthesis, Syncore® Polyvap, Syncore® Analyst
Glass sample tubes for rack R-12 polyvap, round bottom (Ø 48 mm), height 170 mm, for working volume up to 120 ml
Price
516,00 €
Log in for availability
1 * 12 ST
Product Details & Documents
Specifications
  • Description:
    Glass sample tubes for rack R-12 polyvap, round bottom (Ø 48 mm), height 170 mm, for working volume up to 120 ml
  • Seq:
    05
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.