Dropper tips, WHEATON®
Grey
Catalog # WHEAW242425-A
Supplier: DWK Life Sciences
Aller à :
Product Details & Documents
LDPE, coloured.
- Extended controlled dropper tips
- Dispense one drop at a time
- Average drop size is 44 ±6 μl
Ordering information: Tips available in different colours and packaged in case of 100 or 1000.
Spécifications
- Colour:Grey
- Seq:50