Orders LinkContactUsLinkComponent
Dropper tips, WHEATON®
Red
  215-4813
Dropper tips, WHEATON®
  215-4813
Dropper tips, WHEATON®
Dropper tips, WHEATON®
Red
296,00 €
1 * 1.000 ST
LDPE, coloured.
  • Extended controlled dropper tips
  • Dispense one drop at a time
  • Average drop size is 44 ±6 μl
 : Tips available in different colours and packaged in case of 100 or 1000.
  • Colour:
    Red
  • Seq:
    06