Orders LinkContactUsLinkComponent
Dropper tips, WHEATON®
Dropper tips, WHEATON®
Pink
Catalog # 215-4831
Supplier:   DWK Life Sciences
Dropper tips, WHEATON®
Catalog # 215-4831
Supplier:  DWK Life Sciences
Dropper tips, WHEATON®
Dropper tips, WHEATON®
Pink
Price
436,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
LDPE, coloured.
  • Extended controlled dropper tips
  • Dispense one drop at a time
  • Average drop size is 44 ±6 μl
Ordering information: Tips available in different colours and packaged in case of 100 or 1000.
Specifications
  • Colour:
    Pink
  • Seq:
    63