Dropper tips, WHEATON®
Green
Catalog # 215-4835
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
LDPE, coloured.
- Extended controlled dropper tips
- Dispense one drop at a time
- Average drop size is 44 ±6 μl
Informations de commande: Tips available in different colours and packaged in case of 100 or 1000.
Specifications
- Colour:Green
- Seq:71