Orders LinkContactUsLinkComponent
Trypsinizing flasks, WHEATON®
Trypsinizing flasks, WHEATON®
300 ml, Top screw cap 38-430, Screw cap 20-415
Catalog # WHEA355755
Supplier:   DWK Life Sciences
Trypsinizing flasks, WHEATON®
Catalog # WHEA355755
Supplier:  DWK Life Sciences
Trypsinizing flasks, WHEATON®
Trypsinizing flasks, WHEATON®
300 ml, Top screw cap 38-430, Screw cap 20-415
Price
175,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Borosilicate glass, clear.
  • With pour out and caps
  • Fluted design improves mixing
  • Deep skirted screw caps
These flasks feature deep-fluted sides to promote mixing. A dam below the sidearm retains untrypsinized tissue when decanting. A deep skirted screw cap on the sidearm prevents contamination of flask contents. The sidearm allows easier decanting of contents while retaining large particles behind the dam. Sidearm caps are 20-415 size. Caps have white styrene-butadiene rubber liners.
Specifications
  • Capacity:
    300 ml
  • Description:
    Top screw cap 38-430
  • Neck:
    Screw cap 20-415