Orders LinkContactUsLinkComponent
Trypsinizing flasks, WHEATON®
Trypsinizing flasks, WHEATON®
35 ml, Top screw cap 24-430
Catalog # 392-0369
Supplier:   DWK Life Sciences
Trypsinizing flasks, WHEATON®
Catalog # 392-0369
Supplier:  DWK Life Sciences
Trypsinizing flasks, WHEATON®
Trypsinizing flasks, WHEATON®
35 ml, Top screw cap 24-430
Price
144,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Borosilicate glass, clear.
  • With pour out and caps
  • Fluted design improves mixing
  • Deep skirted screw caps
These flasks feature deep-fluted sides to promote mixing. A dam below the sidearm retains untrypsinized tissue when decanting. A deep skirted screw cap on the sidearm prevents contamination of flask contents. The sidearm allows easier decanting of contents while retaining large particles behind the dam. Sidearm caps are 20-415 size. Caps have white styrene-butadiene rubber liners.
Specifications
  • Capacity:
    35 ml
  • Description:
    Top screw cap 24-430
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.