Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Scintillation vials, WHEATON®

20,00 ml, 27×61 mm, Yes, Separate, 22 mm, PE
  WHEA986714
Scintillation vials, WHEATON®
  WHEA986714
Scintillation vials, WHEATON®
Scintillation vials, WHEATON®

20,00 ml, 27×61 mm, Yes, Separate, 22 mm, PE

 
332,00 €
   
1 * 500 ST
 

These scintillation vials made from HDPE are available with screw cap attached or separately in PP or PE and different liner material depending on the application.

 
  • Cap included:
    Yes
  • Cap material:
    PE
  • Cap Size:
    22 mm
  • Ø×H:
    27×61 mm
  • Capacity:
    20,00 ml
  • Seq:
    10
  • Cap type:
    Separate