Orders LinkContactUsLinkComponent
Dropper tips, WHEATON®
Yellow
  215-4783
Dropper tips, WHEATON®
  215-4783
Dropper tips, WHEATON®
Dropper tips, WHEATON®
Yellow
34,00 €
1 * 100 ST
LDPE, coloured.
  • Extended controlled dropper tips
  • Dispense one drop at a time
  • Average drop size is 44 ±6 μl
 : Tips available in different colours and packaged in case of 100 or 1000.
  • Colour:
    Yellow
  • Seq:
    17