Order Entry
Belgium
Orders LinkContactUsLinkComponent
Filling solutions for combined/reference electrodes

Filling solutions for combined/reference electrodes

KCl solution, saturated, without AgCl, for appropriate Red Rod & Calomel reference elements, 100 ml bottle, 100 mL
Catalog # HACHS21M002
Supplier:   Hach
Filling solutions for combined/reference electrodes
Catalog # HACHS21M002
Supplier:  Hach
CAS Number:  
Filling solutions for combined/reference electrodes
Filling solutions for combined/reference electrodes

KCl solution, saturated, without AgCl, for appropriate Red Rod & Calomel reference elements, 100 ml bottle, 100 mL

CAS Number:  
Price
25,90 €
Log in for availability
1 * 100 mL
Product Details & Documents

Filling and storage solutions for combined and reference electrodes.

Documents

Specifications

Specifications

  • Pk:
    100 mL
  • Description:
    KCl solution, saturated, without AgCl, for appropriate Red Rod & Calomel reference elements, 100 ml bottle

Specifications

Aanbevelingen

Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.

Anders ontvangt u algemene aanbevelingen.