Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Certified colour reference standards

ASTM Colour 1, (certified to ISO 17025), 500 mL
Catalog # 633-0004
Supplier:   Lovibond Tintometer
Certified colour reference standards
Catalog # 633-0004
CAS Number:  
Certified colour reference standards
Certified colour reference standards

ASTM Colour 1, (certified to ISO 17025), 500 mL

CAS Number:  
Price
266,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

For routine calibration of colorimeters and verification of test data.

Lovibond® certified colour reference materials are supplied with full traceability to internationally recognised standards. Each standard comes in a 500 ml bottle with a 12-month guarantee of colour stability and full certification including MSDS. Individual values may vary but these are always specified on the certificate of calibration supplied with each standard.

 

Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    ASTM Colour 1, (certified to ISO 17025)

Specifications