Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tissue processing/embedding cassettes, Histosette® II, M485

Tissue processing/embedding cassettes, Histosette® II, M485

Beige
Catalog # 720-0232
Supplier:   Simport Scientific
Tissue processing/embedding cassettes, Histosette® II, M485
Catalog # 720-0232
Supplier:  Simport Scientific
Tissue processing/embedding cassettes, Histosette® II, M485
Tissue processing/embedding cassettes, Histosette® II, M485

Beige

Price
123,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

Acetal polymer, similar to M492, but covers are already removed from cassettes and are packed separately in the case.

  • Convenient for cassette labelling instruments as the covers are already removed
  • Resistant to the chemical action of histological solvents
  • Flow-through slots maximise fluid exchange
Specifications
  • Colour:
    Beige
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.