Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tissue processing/embedding cassettes, Histosette® II, M485

Tissue processing/embedding cassettes, Histosette® II, M485

Salmon
Catalog # 720-0231
Supplier:   Simport Scientific
Tissue processing/embedding cassettes, Histosette® II, M485
Catalog # 720-0231
Supplier:  Simport Scientific
Tissue processing/embedding cassettes, Histosette® II, M485
Tissue processing/embedding cassettes, Histosette® II, M485

Salmon

Price
123,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

Acetal polymer, similar to M492, but covers are already removed from cassettes and are packed separately in the case.

  • Convenient for cassette labelling instruments as the covers are already removed
  • Resistant to the chemical action of histological solvents
  • Flow-through slots maximise fluid exchange
Specifications
  • Colour:
    Salmon