Sample containers, with screw cap
10 ml, 21,0 mm, 58,4 mm
216-3104
: KARTELL
To store and transport samples securely.
LDPE, translucent.
- Good chemical resistance, good temperature resistance
- Strong walls, slightly conical shape
- Particularly tight seal through screw cap with cone gasket
- Ext.Ø:21,0 mm
- Capacity:10 ml
- Height:58,4 mm
- Seq:02



