Assortment Boxes, PS
Assortment box, with 10 compartments, Transparent, 170×105×32 mm, PS
Catalog # HUEN617300
Supplier: hünersdorff GmbH
Jump to:
Product Details & Documents
These assortment boxes made of high impact strength polystyrene (PS) are suitable for packaging purposes and create order in workshops and in household.
- Available either with versatile compartment subdivisions and versions or without compartments
- Transparent lid and bottom for optimum display of the contents or in colours suitable for foods
- Printing possible on request
Specifications
- Colour:Transparent
- Description:Assortment box, with 10 compartments
- Material:PS
- Seq:01
- W×D×H:170×105×32 mm