Assortment Boxes, PS
Assortment box, with 18 compartments, Transparent, 210×120×36 mm, PS
Catalog # HUEN616400
Supplier: hünersdorff GmbH
Jump to:
Product Details & Documents
These assortment boxes made of high impact strength polystyrene (PS) are suitable for packaging purposes and create order in workshops and in household.
- Available either with versatile compartment subdivisions and versions or without compartments
- Transparent lid and bottom for optimum display of the contents or in colours suitable for foods
- Printing possible on request
Specifications
- Colour:Transparent
- Description:Assortment box, with 18 compartments
- Material:PS
- Seq:05
- W×D×H:210×120×36 mm
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.