Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH electrode solutions

pH electrode solutions

KCl reference electrolyte, 3 M/L, 250 ml, 250 mL
Catalog # 662-0282
Supplier:   WTW
pH electrode solutions
Catalog # 662-0282
Supplier:  WTW
CAS Number:  
pH electrode solutions
pH electrode solutions

KCl reference electrolyte, 3 M/L, 250 ml, 250 mL

CAS Number:  
Price
31,80 €
Log in for availability
1 * 250 mL
Product Details & Documents

Solutions for operation, cleaning and storage of electrodes - simple refilling and safe storage warranted.

Electrolytes are used as refill solutions for liquid filled electrodes as well as a storage solution for all types of pH electrodes. With the pepsin cleaning solution, you can remove protein precipitation from the diaphragm of electrodes with liquid electrolyte.

Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    KCl reference electrolyte, 3 M/L, 250 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.