Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Bone curettes, Aesculap®

Bone curette, type Simon (Fig. 00), with 4,4 mm end, 235 mm
  AESCFK689R
 :   Aesculap
Bone curettes, Aesculap®
  AESCFK689R
 :  Aesculap
Bone curettes, Aesculap®

Bone curette, type Simon (Fig. 00), with 4,4 mm end, 235 mm

 
 
   
1 * 1 ST
 
 
  • Description:
    Bone curette, type Simon (Fig. 00), with 4,4 mm end
  • Length:
    235 mm
  • Seq:
    43