Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Bone curettes, Aesculap®

Bone curette, Schede type (Fig. 4), with 8,0 mm end, 170 mm
Catalog # AESCFK627R
Supplier:   Aesculap
Bone curettes, Aesculap®
Catalog # AESCFK627R
Supplier:  Aesculap
Bone curettes, Aesculap®

Bone curette, Schede type (Fig. 4), with 8,0 mm end, 170 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Bone curette, Schede type (Fig. 4), with 8,0 mm end
  • Length:
    170 mm
  • Seq:
    14