Order Entry
Belgium
Orders LinkContactUsLinkComponent
Bone curettes, Aesculap®

Bone curettes, Aesculap®

Bone curette, Schede type (Fig. 3), with 7,4 mm end, 170 mm
Catalog # AESCFK626R
Supplier:   Aesculap
Bone curettes, Aesculap®
Catalog # AESCFK626R
Supplier:  Aesculap
Bone curettes, Aesculap®

Bone curette, Schede type (Fig. 3), with 7,4 mm end, 170 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Bone curette, Schede type (Fig. 3), with 7,4 mm end
  • Length:
    170 mm
  • Seq:
    13
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.