Orders LinkContactUsLinkComponent
Dressing forceps, Aesculap®
Straight, blunt, serrated, 115 mm
  AESCBD043R
 :   Aesculap
Dressing forceps, Aesculap®
  AESCBD043R
 :  Aesculap
Dressing forceps, Aesculap®
Straight, blunt, serrated, 115 mm
24,10 €
1 * 1 ST
These dissecting forceps are ideal for dental surgery.
  • Straight
  • Serrated tips
  • Type:
    Straight, blunt, serrated
  • Length:
    115 mm
  • Seq:
    02