Order Entry
Belgium
Orders LinkContactUsLinkComponent
Double spatulas, flexible

Double spatulas, flexible

250 mm, 60 mm, 20 mm
Catalog # BOCH3644
Supplier:   BOCHEM
Double spatulas, flexible
Catalog # BOCH3644
Supplier:  BOCHEM
Double spatulas, flexible
Double spatulas, flexible

250 mm, 60 mm, 20 mm

Price
7,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel 18/10.

Specifications
  • Length:
    250 mm
  • Blade width:
    20 mm
  • Blade length:
    60 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.