Order Entry
Belgium
Orders LinkContactUsLinkComponent
Double spatulas, flexible

Double spatulas, flexible

210 mm, 52 mm, 20 mm
Catalog # BOCH3643
Supplier:   BOCHEM
Double spatulas, flexible
Catalog # BOCH3643
Supplier:  BOCHEM
Double spatulas, flexible
Double spatulas, flexible

210 mm, 52 mm, 20 mm

Price
6,50 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel 18/10.

Specifications
  • Length:
    210 mm
  • Blade width:
    20 mm
  • Blade length:
    52 mm