Order Entry
Belgium
Orders LinkContactUsLinkComponent
Wire baskets, square and rectangular

Wire baskets, square and rectangular

180×180×180 mm
Catalog # BOCH10029
Supplier:   BOCHEM
Wire baskets, square and rectangular
Catalog # BOCH10029
Supplier:  BOCHEM
Wire baskets, square and rectangular
Wire baskets, square and rectangular

180×180×180 mm

Price
83,80 €
Log in for availability
1 * 1 ST
Product Details & Documents

These stainless steel baskets are designed for transporting, storing, cleaning and sterilising laboratory utensils.

  • Spot-welded grid 8×8×1 mm
  • Reinforced edges
  • Very good chemical and temperature resistance
Specifications
  • W×D×H:
    180×180×180 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.