Order Entry
Belgium
Orders LinkContactUsLinkComponent
Retort rods, without thread

Retort rods, without thread

12 mm, 500 mm
Catalog # BOCH5160
Supplier:   BOCHEM
Retort rods, without thread
Catalog # BOCH5160
Supplier:  BOCHEM
Retort rods, without thread
Retort rods, without thread

12 mm, 500 mm

Price
16,20 €
Log in for availability
1 * 1 ST
Product Details & Documents

Galvanised steel.

Specifications
  • Ø:
    12 mm
  • Length:
    500 mm