Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Spoons, POLY

Oblique, 500 mm, 65×30 mm
  231-0605
 :   BOCHEM
Spoons, POLY
  231-0605
 :  BOCHEM
Spoons, POLY
Spoons, POLY

Oblique, 500 mm, 65×30 mm

 
17,10 €
   
1 * 1 ST
 

Stainless steel 18/10, anti-magnetic. Multipurpose spoon for a wide range of uses. Special shape for light and easy handling.

  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean
 
  • Description:
    Oblique
  • Length:
    500 mm
  • Seq:
    11
  • Spoon size:
    65×30 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.