Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Stand bases

1,8 kg, 130×210×8 mm
Catalog # BOCH5035
Supplier:   BOCHEM
Stand bases
Catalog # BOCH5035
Supplier:  BOCHEM
Stand bases
Stand bases

1,8 kg, 130×210×8 mm

Price
46,70 €
Log in for availability
1 * 1 ST
Product Details & Documents

Steel, varnished.

  • With M10 threaded hole
  • One adjustable foot

 : DIN 12892

Specifications
  • W×D×H:
    130×210×8 mm
  • Weight:
    1,8 kg