Order Entry
Belgium
Orders LinkContactUsLinkComponent
Retort rods, with M10 thread

Retort rods, with M10 thread

12 mm, 1500 mm, M10
Catalog # BOCH5115
Supplier:   BOCHEM
Retort rods, with M10 thread
Catalog # BOCH5115
Supplier:  BOCHEM
Retort rods, with M10 thread
Retort rods, with M10 thread

12 mm, 1500 mm, M10

Price
54,40 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel 18/10.

  • Rods for stand bases
  • M10 thread
Specifications
  • Ø:
    12 mm
  • Thread:
    M10
  • Length:
    1500 mm
  • Seq:
    06
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.