Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sockets for rods without thread

Sockets for rods without thread

13,2 mm, Socket without thread, green coated, for 12 - 13 mm Ø rods, Steel
Catalog # BOCH5044
Supplier:   BOCHEM
Sockets for rods without thread
Catalog # BOCH5044
Supplier:  BOCHEM
Sockets for rods without thread
Sockets for rods without thread

13,2 mm, Socket without thread, green coated, for 12 - 13 mm Ø rods, Steel

Price
14,90 €
Log in for availability
1 * 1 ST
Product Details & Documents

These sockets are available in two materials: 18/10 stainless steel or green coated steel.

Suitable for rods with a diameter of 12 - 13 mm.

Specifications
  • Ø:
    13,2 mm
  • Description:
    Socket without thread, green coated, for 12 - 13 mm Ø rods
  • Material:
    Steel
  • Seq:
    03