Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Retort rods, without thread

16 mm, 1000 mm
Catalog # BOCH5126
Supplier:   BOCHEM
Retort rods, without thread
Catalog # BOCH5126
Supplier:  BOCHEM
Retort rods, without thread
Retort rods, without thread

16 mm, 1000 mm

Price
69,20 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel 18/10.

  • Rods for stand bases
  • Without thread
Specifications
  • Ø:
    16 mm
  • Length:
    1000 mm
  • Seq:
    16