Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Diffraction/transmission gratings
Diffraction/transmission gratings
Transmission grating 300 lines/mm
Catalog # BINH1003080
Marketsource
Supplier:   3B Scientific
Diffraction/transmission gratings
Catalog # BINH1003080
Supplier:  3B Scientific
Marketsource
Diffraction/transmission gratings
Diffraction/transmission gratings
Transmission grating 300 lines/mm
Price
104,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Transmission grating for spectroscopic examinations and for experiments on diffraction and interference. Suitable to resolve the sodium D lines.
  • Mounted on glass carrier
  • Dimensions: 38×50 mm
Specifications
  • Désignation:
    Transmission grating 300 lines/mm