Diffraction/transmission gratings
Transmission grating 600 lines/mm
Catalog # BINH1003079
Marketsource
Supplier: 3B Scientific
Jump to:
Product Details & Documents
Transmission grating for spectroscopic examinations and for experiments on diffraction and interference. Suitable to resolve the sodium D lines.
- Mounted on glass carrier
- Dimensions: 38×50 mm
Specifications
- Description:Transmission grating 600 lines/mm