Orders LinkContactUsLinkComponent
Dissecting scissors, OP-SPECIAL
Dissecting scissors, OP-SPECIAL
Curved, OP-special, extra sharp/sharp, 105 mm
Catalog # HAMML-HSB391-10
Supplier:   HAMMACHER KARL
Dissecting scissors, OP-SPECIAL
Catalog # HAMML-HSB391-10
Supplier:  HAMMACHER KARL
Dissecting scissors, OP-SPECIAL
Dissecting scissors, OP-SPECIAL
Curved, OP-special, extra sharp/sharp, 105 mm
Price
115,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Extra sharp dissection scissors for surgical operations.
Specifications
  • Type:
    Curved, OP-special, extra sharp/sharp
  • Length:
    105 mm
  • Seq:
    02