Docusate sodium ≥96%
Plastic bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 96% CAS No.: 577-11-7 Molecular Formula: C20H37NaO7S Formula Weight: 444.55 Physical Form: Solid Density: 1.1 Melting Point: 153-157° Solubility: Soluble in water, acetone, alcohol Merck Reference: 14,3401 MDL No.: MFCD00012455
Wetting and solubilizing agent
Synonyms: Dioctyl sulphosuccinate sodium salt, Dioctyl sulphosuccinate, sodium salt, Sodium dioctyl sulphosuccinate, Sodium 1,4-bis((2-ethylhexyl)oxy)-1,4-dioxobutane-2-sulphonate, bis(2-Ethylhexyl) sodium sulphosuccinate
- CAS Number:577-11-7
- Formula:C₂₀H₃₇NaO₇S
- MDL Number:MFCD00012455
- Molecular Weight:444.57 g/mol
- Merck Index:14,3401
- Boiling Pt:∼180 °C (1013 hPa)
- Melting Pt:153…157 °C
- Density:1.1
- Storage Temperature:Ambient
- EINECS:209-406-4
Documents
Specifications
Specifications
- Pack type:Plastic bottle for solids
- Pk:100 g



