Tissue forceps
Tissue forceps 1:2, 115 mm
Catalog # 232-0001
Supplier: BOCHEM
Jump to:
Product Details & Documents
Stainless steel.
- With small teeth (1:2) at the end of the blades
- Designed for handling tissues with minimal trauma during surgery
Documents
Specifications
- Type:Tissue forceps 1:2
- Length:115 mm