Orders LinkContactUsLinkComponent
Rectangular cover glasses, Menzel Gläser
Rectangular cover glasses, Menzel Gläser
Cover glasses, 24×50 mm, #0
Catalog # 630-2868
Supplier:   Epredia
Rectangular cover glasses, Menzel Gläser
Catalog # 630-2868
Supplier:  Epredia
Rectangular cover glasses, Menzel Gläser
Rectangular cover glasses, Menzel Gläser
Cover glasses, 24×50 mm, #0
Price
97,50 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
These cover slips are suitable for in vitro diagnostic (IVD) applications.
  • Uniform quality of thickness
  • No bubbles or flaws
  • Made of pre-washed pure white Hydrolytic Class 1 glass
Documents
Specifications
  • No.:
    #0
  • Description:
    Cover glasses
  • L×W:
    24×50 mm
  • Seq:
    25