Orders LinkContactUsLinkComponent
Slide mailers
Slide mailers
Slide mailers, Translucent, PP, 84×100×6 mm, 3
Catalog # 631-9161
Supplier:   KARTELL
Slide mailers
Catalog # 631-9161
Supplier:  KARTELL
Slide mailers
Slide mailers
Slide mailers, Translucent, PP, 84×100×6 mm, 3
Price
39,70 €
Log in for availability
1 * 10 ST
Product Details & Documents
These mailers are suitable for the safe transportation of cytological and geological microscope slides.
  • Separate compartment with guide rails and more space for each slide
  • Flush closing
  • Lightweight and flat
W×D×H: 50×100×6 mm (for 1); 67×85×6 mm (for 2); 84×100×6 mm (for 3)
Specifications
  • No. of slides:
    3
  • Colour:
    Translucent
  • Description:
    Slide mailers
  • Material:
    PP
  • Seq:
    003
  • W×D×H:
    84×100×6 mm