Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sample containers, with screw cap

Sample containers, with screw cap

30 ml, 35,0 mm, 53,0 mm
  216-3106
 :   KARTELL
Sample containers, with screw cap
  216-3106
 :  KARTELL
Sample containers, with screw cap
Sample containers, with screw cap

30 ml, 35,0 mm, 53,0 mm

 
64,20 €
   
1 * 10 ST
 

To store and transport samples securely.
LDPE, translucent.

  • Good chemical resistance, good temperature resistance
  • Strong walls, slightly conical shape
  • Particularly tight seal through screw cap with cone gasket
 
  • Ext.Ø:
    35,0 mm
  • Capacity:
    30 ml
  • Height:
    53,0 mm
  • Seq:
    03