Order Entry
Belgium
Orders LinkContactUsLinkComponent
n-Pentane ≥95%, TECHNICAL

n-Pentane ≥95%, TECHNICAL

Aluminium bottle, 5 L
Catalog # 26192.361
Flammable
Health
Irritant
Environment
Supplier:   VWR Chemicals
n-Pentane ≥95%, TECHNICAL
Catalog # 26192.361
Supplier:  VWR Chemicals
CAS Number:  109-66-0
n-Pentane ≥95%, TECHNICAL
n-Pentane ≥95%, TECHNICAL

Aluminium bottle, 5 L

CAS Number:  109-66-0
Price
138,00 €
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Pentane
  • CAS Number:109-66-0
  • Formula:H₃C(CH₂)₃CH₃
  • MDL Number:MFCD00009498
  • Molecular Weight:72.15 g/mol
  • Merck Index:13,07193
  • Boiling Pt:35…36 °C (1013 hPa)
  • Melting Pt:–130 °C
  • Density:0.63 g/cm³ (20 °C)
  • Flash Pt:–40 °C
  • Storage Temperature:Ambient
  • EINECS:203-692-4
  • UN:1265
  • ADR:3,II
  • REACH:01-2119459286-30
Specifications

Specification Test Results

  • Assay:
    Min. 95 %
  • Boiling point:
    35 - 37 °C

Specifications

  • Pack type:
    Aluminium bottle
  • Pk:
    5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.