Ethyl acetate ≥99%, TECHNICAL
Plastic drum, 25 L
Supplier: VWR Chemicals
Ethyl acetate ≥99%, TECHNICAL
Catalog # 23879.466
Supplier: VWR Chemicals
CAS Number: 141-78-6
Jump to:
Product Details & Documents
Synonyms: Acetic acid ethyl ester
- CAS Number:141-78-6
- Formula:CH₃COOC₂H₅
- MDL Number:MFCD00009171
- Molecular Weight:88.11 g/mol
- Merck Index:13,03792
- Boiling Pt:77.1 °C (1013 hPa)
- Melting Pt:–83 °C
- Density:0.902 g/cm³ (20 °C)
- Flash Pt:–4 °C
- Storage Temperature:Ambient
- EINECS:205-500-4
- UN:1173
- ADR:3,II
- REACH:01-2119475103-46
Documents
Certificates
Search by Lot Number1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 225L Metal drum UN (ZPACE8010050) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic jerrycan for liquids DIN 42 (ZPACK13951025)
Packaging Information
P20_VWRC23879.295_SPEC_EN.pdf P20_VWRC23879.364_SPEC_EN.pdf P20_VWRC23879.466_SPEC_EN.pdf P20_VWRC23879.557_SPEC_EN.pdf spec_VWRC23879.295
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Specifications
Specification Test Results
- Assay:Min. 99 %
- Appearance:Clear colourless liquid
- IR Spectrum:Passes test
Specifications
- Pack type:Plastic drum
- Pk:25 L
- Cat. No.:23879.466
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
- Environmentally Preferable:Safer Human and Environmental Health
Specifications
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.



