Orders LinkContactUsLinkComponent
Robotic tips for Tecan®
Robotic tips for Tecan®
904-251G (wide bore), 1000 µl, +, -, Black, 10 racks, 96 each
Catalog # 736-0062
   Low Manufacturing Impact
Robotic tips for Tecan®
Catalog # 736-0062
   Low Manufacturing Impact
Robotic tips for Tecan®
Robotic tips for Tecan®
904-251G (wide bore), 1000 µl, +, -, Black, 10 racks, 96 each
Price
224,00 €
Log in for availability
1 * 960 ST
Product Details & Documents
High quality tips, especially intended for research in molecular biology laboratories.
  • Transparent or black PP
  • Tips with ART® filters: Eliminate any risk of cross-contamination between samples; designed for applications with DNA, RNA as well as radioactive and infectious samples
  • Certified free of RNase, DNase, DNA and pyrogens
  • Sterile tips: Sterilised by gamma radiation
  • Wide aperture tips: Designed for applications with more viscous samples
Ordering information: Available in boxes of 10 racks or 50 racks (Econopak).
Specifications
  • Packed:
    10 racks, 96 each
  • Sterile:
    +
  • Volume:
    1000 µl
  • Tip style:
    904-251G (wide bore)
  • Tip colour:
    Black
  • Filter:
    -
  • Cat. No.:
    736-0062
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Environmentally Preferable:
    Low Manufacturing Impact