Order Entry
Belgium
Orders LinkContactUsLinkComponent
Filling solutions for combined/reference electrodes

Filling solutions for combined/reference electrodes

KCl solution, saturated, 30 ml bottle, 30 mL
Catalog # HACHS21M010
Supplier:   Hach
Filling solutions for combined/reference electrodes
Catalog # HACHS21M010
Supplier:  Hach
CAS Number:  
Filling solutions for combined/reference electrodes
Filling solutions for combined/reference electrodes

KCl solution, saturated, 30 ml bottle, 30 mL

CAS Number:  
Price
18,20 €
Log in for availability
1 * 30 mL
Product Details & Documents

Filling and storage solutions for combined and reference electrodes.

Documents

Specifications

Specifications

  • Pk:
    30 mL
  • Description:
    KCl solution, saturated, 30 ml bottle

Specifications