Filling and storage solution for combined/reference electrodes
Filling and storage solution for combined/reference electrodes, K₂SO₄, 500 ml bottle, 500 mL
Catalog # HACHC20C500
Supplier: Hach
Filling and storage solution for combined/reference electrodes
Catalog # HACHC20C500
Supplier: Hach
CAS Number:
Jump to:
Product Details & Documents
Shelf life: 48 months from production date.
Specifications
Specifications
- Pk:500 mL
- Description:Filling and storage solution for combined/reference electrodes, K₂SO₄, 500 ml bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



