Comparator kits, Lovibond® system 2000
Platinum-Cobalt Units (10 - 250 mg Pt/l) Nessleriser 2150 with Daylight 2000 Lighting unit and 113 mm (50 ml) Nessler cylinders; Pt-Co discs NSH (10 - 90 mg Pt/l), NSB (70 - 250 mg Pt/l); stand for using Nessleriser with natural lighting, AF325
Catalog # 633-0021
Supplier: Lovibond Tintometer
Jump to:
Product Details & Documents
Available for many of the commonly used colour scales, these kits are a convenient means of ordering the complete range of equipment required. Each system includes.
- A visual comparator instrument which brings the sample and colour standards into the same field of view for more accurate colour matching
- A selection of colour standards - stable-coloured glass filters in test discs, which have been pre-calibrated to give direct readings over set measuring ranges
- A standardised benchtop light source to ensure constant, uniform lighting conditions for colour matching and, therefore, consistent and accurate readings
- Glassware of the appropriate type and path length
Specifications
- Description:Platinum-Cobalt Units (10 - 250 mg Pt/l) Nessleriser 2150 with Daylight 2000 Lighting unit and 113 mm (50 ml) Nessler cylinders; Pt-Co discs NSH (10 - 90 mg Pt/l), NSB (70 - 250 mg Pt/l); stand for using Nessleriser with natural lighting
- Type:AF325
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.