Order Entry
Belgium
Orders LinkContactUsLinkComponent
Staining jars

Staining jars

Cylindrical staining jar with cover, - mm, - mm, 40 mm, 85 mm
Catalog # 631-0349
Staining jars
Catalog # 631-0349
Staining jars
Staining jars

Cylindrical staining jar with cover, - mm, - mm, 40 mm, 85 mm

Price
15,10 €
Log in for availability
1 * 1 ST
Product Details & Documents

Soda-lime glass.

These staining jars have an overlapping cover.

Specifications
  • Ø:
    40 mm
  • Width:
    - mm
  • Description:
    Cylindrical staining jar with cover
  • Height:
    85 mm
  • W×D×H:
    - mm