Microplates, 96-well
96-well microplate, V-bottom, MICROLON® 600, 40 - 200 µl, 234 µl
738-0009
Crystal clear PS, without lid. Ideal for ELISA methodologies, these plates are available with high binding surfaces that display a higher protein binding capacity for polar proteins (MICROLON® 600) or medium binding surfaces, which are more hydrophobic and are therefore better suited for non-polar proteins and peptides (MICROLON® 200).
- Available with a choice of well profile: U-bottom, V-bottom, F-bottom (standard), F-bottom (chimney well), or half area
- Free of DNase, RNase and human DNA, and are non-pyrogenic
Documents
- Description:96-well microplate, V-bottom, MICROLON® 600
- Seq:13
- Working volume:40 - 200 µl
- Total volume:234 µl
- Cat. No.:738-0009
- Environmental certification:ISO 14001 (241002); ISO 50001 (241002)
- Environmentally Preferable:Low Manufacturing Impact
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.