Polyethylene stoppers
White, PE, 19/26
Catalog # AZLOTRF595
Supplier: SciLabware
Jump to:
Product Details & Documents
These PE stoppers are ideally suited for use with glass or plastic volumetric flasks and measuring cylinders etc. which have longer 'B' length sockets.
Large head provides a good grip and gives protection if glassware is accidently knocked over.
Packaging: 10/package
Specifications
- NS:19/26
- Colour:White
- Material:PE