Standard micro Pasteurpipette, 155mm, pk500 1 * 500 items
Catalog # 612-0000
Supplier: Alpha Laboratories
Standard micro Pasteurpipette, 155mm, pk500 1 * 500 items
Catalog # 612-0000
Supplier: Alpha Laboratories
Frequently Bought Together
Sodium hydroxide ≥98%, pellets, EMPROVE® ESSENTIAL Ph. Eur., BP, FCC, JP, NF, E524, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Disodium hydrogen phosphate dodecahydrate, crystallised, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
pH buffer solutions
Supplier: Mettler - Toledo
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Combitips® advanced Eppendorf Quality Positive Displacement Pipette Tips
Supplier: EPPENDORF
2,6-Di-tert-butyl-p-cresol Ph. Eur., NF, E321, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Salicylic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Potassium permanganate 0.05 mol/l (0.25 N) in aqueous solution
Supplier: VWR Chemicals
Molybdic acid ≥85% (MoO₃ basis) ACS
Supplier: Thermo Fisher Scientific
Dichloromethane ≥99.8% (by GC) stabilised, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals