Orders LinkContactUsLinkComponent
Bulk material, Hydromatrix™
Bulk material, Hydromatrix™
Hydromatrix™ bulk material, 1 kg
Catalog # VARI198003
Supplier:   VARIAN
Bulk material, Hydromatrix™
Catalog # VARI198003
Supplier:  VARIAN
CAS Number:  
Bulk material, Hydromatrix™
Hydromatrix™ bulk material, 1 kg
CAS Number:  
Price
332,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
SLE products use a broad-performance, inert diatomaceous earth sorbent for rapid, general sample preparation.
Documents
Specifications
Specifications
  • Pk:
    1 kg
  • Description:
    Hydromatrix™ bulk material